Hydrogen chloride, 2M in diethyl ether
Hydrogen chloride, 2M in diethyl ether
Hydrogen chloride, 2M in diethyl ether
Thermo Scientific Chemicals

Hydrogen chloride, 2M in diethyl ether

CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H26914.AE
also known as H26914-AE
Price (USD)
70.65
Online exclusive
78.10
Save 7.45
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
70.65
Online exclusive
78.10
Save 7.45
Each
Chemical Identifiers
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
FormLiquid
Assay (Aqueous acid-base Titration)1.95-2.15 M
Hydrogen chloride, 2M in diethyl ether is used in organic synthesis to prepare emodin derivatives and polyamine derivatives of betulinic acid and beta-sitosterol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only