(S)-(-)-Indoline-2-carboxylic acid, 97+%
(S)-(-)-Indoline-2-carboxylic acid, 97+%
(S)-(-)-Indoline-2-carboxylic acid, 97+%
Thermo Scientific Chemicals

(S)-(-)-Indoline-2-carboxylic acid, 97+%

CAS: 79815-20-6 | C9H9NO2 | 163.176 g/mol
製品番号(カタログ番号) H27009.06
または、製品番号H27009-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS79815-20-6
IUPAC Name2,3-dihydro-1H-indole-2-carboxylic acid
Molecular FormulaC9H9NO2
InChI KeyQNRXNRGSOJZINA-UHFFFAOYNA-N
SMILESOC(=O)C1CC2=CC=CC=C2N1
さらに表示
Assay (Aqueous acid-base Titration)≥97.0 to ≤103.0%
Assay (Silylated GC)≥97.0%
Appearance (Color)Pale cream to cream to pale brown
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only