3-(Boc-amino)benzeneboronic acid pinacol ester, 97%
3-(Boc-amino)benzeneboronic acid pinacol ester, 97%
3-(Boc-amino)benzeneboronic acid pinacol ester, 97%
Thermo Scientific Chemicals

3-(Boc-amino)benzeneboronic acid pinacol ester, 97%

CAS: 330793-09-4 | C17H26BNO4 | 319.208 g/mol
製品番号(カタログ番号) H27026.03
または、製品番号H27026-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS330793-09-4
IUPAC Nametert-butyl N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
Molecular FormulaC17H26BNO4
InChI KeyANQAOGOIWVMGCH-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NC1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1
さらに表示
FormPowder or crystalline powder
Assay (HPLC)≥96.0%
Melting Point (clear melt)116-121?C
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only