(+/-)-Piperazine-2-carboxylic acid dihydrochloride, 98%
(+/-)-Piperazine-2-carboxylic acid dihydrochloride, 98%
(+/-)-Piperazine-2-carboxylic acid dihydrochloride, 98%
Thermo Scientific Chemicals

(+/-)-Piperazine-2-carboxylic acid dihydrochloride, 98%

CAS: 3022-15-9 | C5H12Cl2N2O2 | 203.063 g/mol
製品番号(カタログ番号) H27101.14
または、製品番号H27101-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3022-15-9
IUPAC Name(3R)-piperazine-1,4-diium-3-carboxylate
Molecular FormulaC5H11N2O2
InChI KeyJSSXHAMIXJGYCS-SCSAIBSYSA-O
SMILES[O-]C(=O)[C@H]1C[NH2+]CC[NH2+]1
さらに表示
Appearance (Color)White to cream to yellow
FormCrystals or powder or crystalline powder or chunks
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only