Appearance (Color)Colourless or white to yellow
FormViscous liquid or fused low melting solid
Assay from Supplier's CofA1-n-Butyl-2,3-dimethylimidazolium cation: ≥98.5% (by Ion Chromatography)
Assay from Supplier's CofATetrafluoroborate anion: ≥98.5% (by Ion Chromatography)
Residual halide≤100ppm (by Ion Chromatography)
さらに表示