1-Butyl-2,3-dimethylimidazolium chloride, 99%
1-Butyl-2,3-dimethylimidazolium chloride, 99%
1-Butyl-2,3-dimethylimidazolium chloride, 99%
1-Butyl-2,3-dimethylimidazolium chloride, 99%
1-Butyl-2,3-dimethylimidazolium chloride, 99%
Thermo Scientific Chemicals

1-Butyl-2,3-dimethylimidazolium chloride, 99%

CAS: 98892-75-2 | C9H17ClN2 | 188.699 g/mol
製品番号(カタログ番号) H27270.06
または、製品番号H27270-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS98892-75-2
IUPAC Name3-butyl-1,2-dimethyl-1H-imidazol-3-ium
Molecular FormulaC9H17N2
InChI KeyXUAXVBUVQVRIIQ-UHFFFAOYSA-N
SMILESCCCC[N+]1=C(C)N(C)C=C1
さらに表示
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥97.0 to ≤103.0%
Water Content (Karl Fischer Titration)≤1.0%
Melting Point (clear melt)93.0-100.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only