1-n-Hexyl-3-methylimidazolium hexafluorophosphate, 99%
1-n-Hexyl-3-methylimidazolium hexafluorophosphate, 99%
1-n-Hexyl-3-methylimidazolium hexafluorophosphate, 99%
1-n-Hexyl-3-methylimidazolium hexafluorophosphate, 99%
Thermo Scientific Chemicals

1-n-Hexyl-3-methylimidazolium hexafluorophosphate, 99%

CAS: 304680-35-1 | C10H19F6N2P | 312.24 g/mol
製品番号(カタログ番号) H27272.18
または、製品番号H27272-18
価格(JPY)
-
見積もりを依頼する
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS304680-35-1
IUPAC Name1-hexyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Molecular FormulaC10H19F6N2P
InChI KeyYPWSSSRXUOQNMQ-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1
さらに表示
Refractive Index1.4165-1.4200 @ 20?C
Appearance (Color)Clear, colorless to pale yellow or yellow to orange
Residual halide<250ppm
Water Content (Karl Fischer Titration)<250ppm
FormLiquid
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only