4-Benzyloxy-2-chloropyrimidine, 95%, Thermo Scientific Chemicals
4-Benzyloxy-2-chloropyrimidine, 95%, Thermo Scientific Chemicals
4-Benzyloxy-2-chloropyrimidine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Benzyloxy-2-chloropyrimidine, 95%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
H27313.03
または、製品番号H27313-03
1 g
製品番号(カタログ番号) H27313.03
または、製品番号H27313-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS108381-28-8
IUPAC Name4-(benzyloxy)-2-chloropyrimidine
Molecular FormulaC11H9ClN2O
InChI KeySNCGVIVLUWJDQN-UHFFFAOYSA-N
SMILESClC1=NC=CC(OCC2=CC=CC=C2)=N1
さらに表示
Assay (GC)≥94.0%
Appearance (Color)White
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only