1-n-Butyl-3-methylimidazolium methanesulfonate, 99%
1-n-Butyl-3-methylimidazolium methanesulfonate, 99%
1-n-Butyl-3-methylimidazolium methanesulfonate, 99%
Thermo Scientific Chemicals

1-n-Butyl-3-methylimidazolium methanesulfonate, 99%

CAS: 342789-81-5 | C9H18N2O3S | 234.314 g/mol
製品番号(カタログ番号) H27336.06
または、製品番号H27336-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS342789-81-5
IUPAC Name1-butyl-3-methyl-1H-imidazol-3-ium methanesulfonate
Molecular FormulaC9H18N2O3S
InChI KeyPUHVBRXUKOGSBC-UHFFFAOYSA-M
SMILESCS([O-])(=O)=O.CCCCN1C=C[N+](C)=C1
さらに表示
Appearance (Color)White
FormCrystals or powder or crystalline powder
Melting Point (clear melt)71.0-77.0?C
Assay (HPLC)≥98.5%
Water Content (Karl Fischer Titration)≤0.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only