3-n-Hexadecylthiophene, 97%
3-n-Hexadecylthiophene, 97%
3-n-Hexadecylthiophene, 97%
Thermo Scientific Chemicals

3-n-Hexadecylthiophene, 97%

CAS: 119269-24-8 | C20H36S | 308.57 g/mol
製品番号(カタログ番号) H27371.06
または、製品番号H27371-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS119269-24-8
IUPAC Name3-hexadecylthiophene
Molecular FormulaC20H36S
InChI KeyFRVZSODZVJPMKO-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCC1=CSC=C1
さらに表示
FormFused solid
Melting Point (clear melt)27.5-36.5?C
Appearance (Color)White
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only