2-Methoxy-4-nitrobenzonitrile, 98%
2-Methoxy-4-nitrobenzonitrile, 98%
2-Methoxy-4-nitrobenzonitrile, 98%
Thermo Scientific Chemicals

2-Methoxy-4-nitrobenzonitrile, 98%

CAS: 101084-96-2 | C8H6N2O3 | 178.147 g/mol
製品番号(カタログ番号) H27391.06
または、製品番号H27391-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS101084-96-2
IUPAC Name2-methoxy-4-nitrobenzonitrile
Molecular FormulaC8H6N2O3
InChI KeyMLIKCKXLGYEGAO-UHFFFAOYSA-N
SMILESCOC1=C(C=CC(=C1)[N+]([O-])=O)C#N
さらに表示
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only