4-Aminobenzeneboronic acid hydrochloride, 97%
4-Aminobenzeneboronic acid hydrochloride, 97%
4-Aminobenzeneboronic acid hydrochloride, 97%
Thermo Scientific Chemicals

4-Aminobenzeneboronic acid hydrochloride, 97%

CAS: 80460-73-7 | C6H9BClNO2 | 173.403 g/mol
数量:
100 mg
500 mg
製品番号(カタログ番号) H27479.MC
または、製品番号H27479-MC
価格(JPY)
-
数量:
100 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS80460-73-7
IUPAC Name(4-aminophenyl)boronic acid
Molecular FormulaC6H8BNO2
InChI KeyMKPDAJWEBQRQCO-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)B(O)O
さらに表示
Appearance (Color)White to pale cream or pale blue
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥95.0%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only