Diphenyl(o-tolyl)phosphine
Diphenyl(o-tolyl)phosphine
Diphenyl(o-tolyl)phosphine
Thermo Scientific Chemicals

Diphenyl(o-tolyl)phosphine

CAS: 5931-53-3 | C19H17P | 276.319 g/mol
製品番号(カタログ番号)数量
H27503.03
または、製品番号H27503-03
1 g
製品番号(カタログ番号) H27503.03
または、製品番号H27503-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5931-53-3
IUPAC Name(2-methylphenyl)diphenylphosphane
Molecular FormulaC19H17P
InChI KeyMLBZLJCMHFCTQM-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
FormPowder
Assay (GC)≥95.0%
Appearance (Color)White
Melting Point (clear melt)66.0-72.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only