4-Bromo-2-nitrobenzoic acid, 97%
4-Bromo-2-nitrobenzoic acid, 97%
4-Bromo-2-nitrobenzoic acid, 97%
Thermo Scientific Chemicals

4-Bromo-2-nitrobenzoic acid, 97%

CAS: 99277-71-1 | C7H3BrNO4 | 245.01 g/mol
製品番号(カタログ番号) H27541.06
または、製品番号H27541-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS99277-71-1
IUPAC Name4-bromo-2-nitrobenzoate
Molecular FormulaC7H3BrNO4
InChI KeyZIRHHEZLJGORGU-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC=C(Br)C=C1[N+]([O-])=O
さらに表示
Appearance (Color)Pale cream
FormPowder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (Silylated GC)≥96.0%
Melting Point (clear melt)163.5-169.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only