Methyl 2-formylthiophene-4-carboxylate, 95%
Methyl 2-formylthiophene-4-carboxylate, 95%
Methyl 2-formylthiophene-4-carboxylate, 95%
Thermo Scientific Chemicals

Methyl 2-formylthiophene-4-carboxylate, 95%

CAS: 67808-66-6 | C7H6O3S | 170.182 g/mol
製品番号(カタログ番号) H27597.03
または、製品番号H27597-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS67808-66-6
IUPAC Namemethyl 5-formylthiophene-3-carboxylate
Molecular FormulaC7H6O3S
InChI KeyGNXNZRYWBFMVHK-UHFFFAOYSA-N
SMILESCOC(=O)C1=CSC(C=O)=C1
さらに表示
Formpowder
Appearance (Color)Brown
Assay (HPLC)> 94.0%
Proton NMRconforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only