Diethyl allyl(3-phenyl-2-propynyl)malonate, 96%, Thermo Scientific Chemicals
Diethyl allyl(3-phenyl-2-propynyl)malonate, 96%, Thermo Scientific Chemicals
Diethyl allyl(3-phenyl-2-propynyl)malonate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl allyl(3-phenyl-2-propynyl)malonate, 96%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
H27702.03
または、製品番号H27702-03
1 g
製品番号(カタログ番号) H27702.03
または、製品番号H27702-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS143633-91-4
IUPAC Name1,3-diethyl 2-(3-phenylprop-2-yn-1-yl)-2-(prop-2-en-1-yl)propanedioate
Molecular FormulaC19H22O4
InChI KeyZSPBIVGRCZUELN-UHFFFAOYSA-N
SMILESCCOC(=O)C(CC=C)(CC#CC1=CC=CC=C1)C(=O)OCC
さらに表示
Appearance (Color)Clear yellow
FormLiquid
Refractive Index1.5110-1.5170 @ 20?C
Assay (GC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only