1,2,2,6,6-Pentamethylpiperidine, 97%
1,2,2,6,6-Pentamethylpiperidine, 97%
1,2,2,6,6-Pentamethylpiperidine, 97%
Thermo Scientific Chemicals

1,2,2,6,6-Pentamethylpiperidine, 97%

CAS: 79-55-0 | C10H21N | 155.285 g/mol
製品番号(カタログ番号) H27729.03
または、製品番号H27729-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS79-55-0
IUPAC Name1,2,2,6,6-pentamethylpiperidin-1-ium
Molecular FormulaC10H22N
InChI KeyXULIXFLCVXWHRF-UHFFFAOYSA-O
SMILESC[NH+]1C(C)(C)CCCC1(C)C
さらに表示
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4570-1.4620 @ 20?C
Identification (FTIR)Conforms
FormLiquid
Assay (GC)≥96.0%
Pharmacological evaluation of the antagonism of nicotine's central effects by mecamylamine and pempidine where pempidine was found to increase the ED50 of nicotine (0.73 mg/kg) for depression of spontaneous activity in a dose-related manner. At a dose of 3 mg/kg, pempidine increased nicotine's ED50 4.7-fold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pharmacological evaluation of the antagonism of nicotine′s central effects by mecamylamine and pempidine where pempidine was found to increase the ED50 of nicotine (0.73 mg/kg) for depression of spontaneous activity in a dose-related manner. At a dose of 3 mg/kg, pempidine increased nicotine′s ED50 4.7-fold.

Solubility
Soluble in toluene, dimethylformamide.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only

General References:

  1. T J Martin,; J Suchocki,; E L May,; B R Martin. Pharmacological evaluation of the antagonism of nicotine's central effects by mecamylamine and pempidine. JPET. 1994, 254 (1), 45-51.
  2. authors. title . Journal name. year of publication , volume issue , pg no.