2,3,4,6-Tetra-O-benzyl-D-glucopyranose, 98+%
2,3,4,6-Tetra-O-benzyl-D-glucopyranose, 98+%
2,3,4,6-Tetra-O-benzyl-D-glucopyranose, 98+%
Thermo Scientific Chemicals

2,3,4,6-Tetra-O-benzyl-D-glucopyranose, 98+%

CAS: 4132-28-9 | C34H36O6 | 540.656 g/mol
製品番号(カタログ番号) H27888.06
または、製品番号H27888-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4132-28-9
IUPAC Name3,4,5-tris(benzyloxy)-6-[(benzyloxy)methyl]oxan-2-ol
Molecular FormulaC34H36O6
InChI KeyOGOMAWHSXRDAKZ-UHFFFAOYNA-N
SMILESOC1OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1
さらに表示
Appearance (Color)White
Assay (HPLC)≥98.0%
Melting Point (clear melt)147.0-153.0?C
Optical Rotation18.0 ± 1.5? (C=2 in chloroform)
FormPowder
It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in chloroform

Notes
Store -20°C. Incompatible with oxidizing agents.
RUO – Research Use Only
Pietro Allevi.; Pierangela Ciuffreda.; Diego Colombo.; Diego Monti.; Giovanna Speranza.; Paolo Manitto. The Wittig-Horner reaction on 2,3,4,6-tetra-O-benzyl-D-mannopyranose and 2,3,4,6-tetra -O-benzyl-D-glucopyranose. J. Chem. Soc., Perkin Trans. 1. 1989, (7),1281-1283 .