Ethyl alpha,4-dibromophenylacetate, 97+%
Ethyl alpha,4-dibromophenylacetate, 97+%
Ethyl alpha,4-dibromophenylacetate, 97+%
Ethyl alpha,4-dibromophenylacetate, 97+%
Thermo Scientific Chemicals

Ethyl alpha,4-dibromophenylacetate, 97+%

CAS: 77143-76-1 | C10H10Br2O2 | 321.996 g/mol
製品番号(カタログ番号) H28093.14
または、製品番号H28093-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS77143-76-1
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥97.0%
Refractive Index1.5675-1.5725 @ 20?C
It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
Achille Cappiello; Giorgio Famiglini; Pierangela Palma; Veronica Termopoli; Helga Trufelli. A new liquid chromatography-mass spectrometry approach for generic screening and quantitation of potential genotoxic alkylation compounds without derivatization. Journal of Chromatography A. 2012, 1255286-290.