Thermo Scientific Chemicals

Methyl thiosalicylate, 97%, Thermo Scientific Chemicals

Catalog number: H28400.14
25 g, Each
Thermo Scientific Chemicals

Methyl thiosalicylate, 97%, Thermo Scientific Chemicals

Catalog number: H28400.14
25 g, Each
Quantity
Catalog number: H28400.14
also known as H28400-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4892-02-8
IUPAC Name
methyl 2-sulfanylbenzoate
Molecular Formula
C8H8O2S
InChI Key
BAQGCWNPCFABAY-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC=C1S
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.5890-1.5940 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text