4-Chloro-6,7-dimethoxyquinazoline, 98%
4-Chloro-6,7-dimethoxyquinazoline, 98%
4-Chloro-6,7-dimethoxyquinazoline, 98%
Thermo Scientific Chemicals

4-Chloro-6,7-dimethoxyquinazoline, 98%

CAS: 13790-39-1 | C10H9ClN2O2 | 224.64 g/mol
製品番号(カタログ番号) H28405.06
または、製品番号H28405-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13790-39-1
IUPAC Name4-chloro-6,7-dimethoxyquinazoline
Molecular FormulaC10H9ClN2O2
InChI KeyLLLHRNQLGUOJHP-UHFFFAOYSA-N
SMILESCOC1=C(OC)C=C2C(Cl)=NC=NC2=C1
さらに表示
Melting Point (clear melt)182.0-188.0?C
Appearance (Color)Pale yellow
FormPowder
Assay (GC)≥97.5%
A useful synthetic intermediate in the preparation of epidermal growth factors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful synthetic intermediate in the preparation of epidermal growth factors.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. R K Narla; X P Liu; D E Myers; and F M Uckun. 4-(3'-Bromo-4'hydroxylphenyl)-amino-6,7-dimethoxyquinazoline: a novel quinazoline derivative with potent cytotoxic activity against human glioblastoma cells. Clin Cancer Res. 1988, 41405.
  2. Chappie, T.A, et.al. Discovery of a Series of 6,7-Dimethoxy-4-pyrrolidylquinazoline PDE10A Inhibitors†.J. Med. Chem. 2007 , 50(2), 182-185.