N-Fmoc-D-proline, 98+%, may cont. up to ca 5% water
N-Fmoc-D-proline, 98+%, may cont. up to ca 5% water
N-Fmoc-D-proline, 98+%, may cont. up to ca 5% water
Thermo Scientific Chemicals

N-Fmoc-D-proline, 98+%, may cont. up to ca 5% water

CAS: 101555-62-8 | C20H19NO4 | 337.375 g/mol
製品番号(カタログ番号) H28500.14
または、製品番号H28500-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS101555-62-8
IUPAC Name1-{[(9H-fluoren-9-yl)methoxy]carbonyl}pyrrolidine-2-carboxylic acid
Molecular FormulaC20H19NO4
InChI KeyZPGDWQNBZYOZTI-UHFFFAOYNA-N
SMILESOC(=O)C1CCCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
さらに表示
FormPowder
Assay (HPLC)≥98.0%
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
In organic synthesis, N-Fmoc-D-proline are often employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis, medicinal chemistry, and polymer chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In organic synthesis, N-Fmoc-D-proline are often employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis, medicinal chemistry, and polymer chemistry.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. asper Kaiser,; Sape S. Kinderman,; Bart C. J. van Esseveldt,; Floris L. van Delft,; Hans E. Schoemaker,; Richard H. Blaauwd,; Floris P. J. T. Rutjes. Synthetic Applications of Aliphatic Unsaturated α-H-α-Amino Acids.. Org. Biomol. Chem. 2005, 33435-3467 .
  2. Proline Derivatives and Analogs.Chemfiles. 512