Thermo Scientific Chemicals

3-Bromopropylboronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog number: H30646.03
1 g, Each
Thermo Scientific Chemicals

3-Bromopropylboronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog number: H30646.03
1 g, Each
Quantity
Catalog number: H30646.03
also known as H30646-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124215-44-7
IUPAC Name
2-(3-bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Formula
C9H18BBrO2
InChI Key
CHQXFJUKMDJWHO-UHFFFAOYSA-N
SMILES
CC1(C)OB(CCCBr)OC1(C)C
Form
Liquid
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.4560-1.4600 @ 20?C

Description

Used as primary and secondary intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as primary and secondary intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text