2,2'-Bis(diphenylphosphino)biphenyl, 98%
2,2'-Bis(diphenylphosphino)biphenyl, 98%
2,2'-Bis(diphenylphosphino)biphenyl, 98%
Thermo Scientific Chemicals

2,2'-Bis(diphenylphosphino)biphenyl, 98%

CAS: 84783-64-2 | C36H28P2 | 522.57 g/mol
数量:
250 mg
1 g
5 g
製品番号(カタログ番号) H31306.06
または、製品番号H31306-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS84783-64-2
IUPAC Name[2'-(diphenylphosphanyl)-[1,1'-biphenyl]-2-yl]diphenylphosphane
Molecular FormulaC36H28P2
InChI KeyGRTJBNJOHNTQBO-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Purity (DSC)>97.5 to <102.5%
Appearance (Color)White
Phosphorus-31 NMRConforms to structure
FormCrystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only