Ethyl 2-cyano-2-methylpropionate, 97%
Ethyl 2-cyano-2-methylpropionate, 97%
Ethyl 2-cyano-2-methylpropionate, 97%
Thermo Scientific Chemicals

Ethyl 2-cyano-2-methylpropionate, 97%

CAS: 1572-98-1 | C7H11NO2 | 141.17 g/mol
製品番号(カタログ番号) H31432.09
または、製品番号H31432-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1572-98-1
IUPAC Nameethyl 2-cyano-2,2-dimethylacetate
Molecular FormulaC7H11NO2
InChI KeyFYGRPGOHQCPZCV-UHFFFAOYSA-N
SMILESCCOC(=O)C(C)(C)C#N
さらに表示
Appearance (Color)Clear colorless to yellow
Refractive Index1.4075-1.4125 @ 20?C
FormLiquid
Assay (GC)≥96.0%
It is employed as a intermediates for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediates for pharmaceutical and organic synthesis.

Solubility
Soluble in ether. Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Hugh W. Thompson; Joseph Swistok. Enamines of 3,3-dimethylazetidine. J. Org. Chem,1981, 46(24), 4907-4911.
  2. Ulhas P. Dhokte; Vien V. Khau; Darrell R. Hutchison; Michael J. Martinelli. A novel approach for total synthesis of cryptophycins via asymmetric crotylboration protocol. Tetrahedron Letters,1998, 39(48), 8771-8774.