2,3-Difluoro-4-hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals
2,3-Difluoro-4-hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals
2,3-Difluoro-4-hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Difluoro-4-hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
H31538.03
または、製品番号H31538-03
1 g
製品番号(カタログ番号) H31538.03
または、製品番号H31538-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS676500-39-3
IUPAC Name2,3-difluoro-4-hydroxybenzaldehyde
Molecular FormulaC7H4F2O2
InChI KeyKTGPDDPEVRJSFY-UHFFFAOYSA-N
SMILESOC1=C(F)C(F)=C(C=O)C=C1
さらに表示
Appearance (Color)Cream or pink
FormPowder
Assay (GC)≥97.5%
Melting Point (clear melt)138.0-145.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only