4-Bromo-2-fluoro-5-(trifluoromethyl)aniline, 97%
4-Bromo-2-fluoro-5-(trifluoromethyl)aniline, 97%
4-Bromo-2-fluoro-5-(trifluoromethyl)aniline, 97%
Thermo Scientific Chemicals

4-Bromo-2-fluoro-5-(trifluoromethyl)aniline, 97%

CAS: 104460-70-0 | C7H4BrF4N | 258.01 g/mol
製品番号(カタログ番号) H31727.09
または、製品番号H31727-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS104460-70-0
Appearance (Color)Pale brown or purple
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)31.0-41.0?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Bromo-2-fluoro-5-(trifluoromethyl)aniline has antimicrobial effect.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. G. J. Fox,; G. Hallas,; J. D. Hepworth,; K. N. Paskins. para-Bromination of Aromatic Amines: 4-Bromo-N,N-dimethyl-3-(trifluoromethyl)aniline. Organic Syntheses.
  2. Bhawani Singh Rathore,; M. Kumar. Synthesis of 7-chloro-5-trifluoromethyl/7-fluoro/7-trifluoromethyl-4H-1,4-benzothiazines as antimicrobial agents. Bioorganic & Medicinal Chemistry. 2006, 14 (16), 5678-5682.