3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals
3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals
3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals

CAS: 1736-21-6 ∣ C11H8FNO3 ∣ 221.19 g/mol
製品番号(カタログ番号) H32090.03
または、製品番号H32090-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1736-21-6
IUPAC Name3-(4-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
Molecular FormulaC11H8FNO3
InChI KeyPDEGBONVUJDOFN-UHFFFAOYSA-N
SMILESCC1=C(C(O)=O)C(=NO1)C1=CC=C(F)C=C1
さらに表示
FormCrystals or crystalline powder
Assay (Silylated GC)≥96.0%
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)197-203?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only