Agarose Beads 4% B-CL (Cross-linked)
Agarose Beads 4% B-CL (Cross-linked)
Agarose Beads 4% B-CL (Cross-linked)
Thermo Scientific Chemicals

Agarose Beads 4% B-CL (Cross-linked)

CAS: 61970-08-9 | C18H19N5O | 321.38 g/mol
製品番号(カタログ番号)数量
H32305.AU
または、製品番号H32305-AU
1000 mL
製品番号(カタログ番号) H32305.AU
または、製品番号H32305-AU
価格(JPY)
-
数量:
1000 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS61970-08-9
IUPAC Name4-{6-[(1-hydroxy-3-methylbutan-2-yl)amino]imidazo[1,2-b]pyridazin-3-yl}benzonitrile
Molecular FormulaC18H19N5O
InChI KeyCQZVJEKKNHOFNB-UHFFFAOYNA-N
SMILESCC(C)C(CO)NC1=NN2C(C=C1)=NC=C2C1=CC=C(C=C1)C#N
さらに表示
Appearance (Color)White
FormSpherical microparticle beads or spherical beads
CommentSuspension in 20% ethanol as antimicrobial agent
Particle SizeVolume share for range 50-150µm: ≥95%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
組成および保存条件
2°C to 8°C