N-Boc-DL-proline, 98%
N-Boc-DL-proline, 98%
N-Boc-DL-proline, 98%
Thermo Scientific Chemicals

N-Boc-DL-proline, 98%

CAS: 59433-50-0 | C10H17NO4 | 215.249 g/mol
製品番号(カタログ番号) H32347.03
または、製品番号H32347-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS59433-50-0
IUPAC Name1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid
Molecular FormulaC10H17NO4
InChI KeyZQEBQGAAWMOMAI-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCCC1C(O)=O
さらに表示
FormCrystalline powder
Proton NMRConforms to structure
Appearance (Color)White
Assay (Aqueous acid-base Titration)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only