1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene, 98%
1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene, 98%
1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene, 98%
Thermo Scientific Chemicals

1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene, 98%

CAS: 286932-57-8 | C7H3BrF4O | 258.998 g/mol
製品番号(カタログ番号) H32650.03
または、製品番号H32650-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS286932-57-8
IUPAC Name2-bromo-1-fluoro-4-(trifluoromethoxy)benzene
Molecular FormulaC7H3BrF4O
InChI KeyVVVFPEJORKPEST-UHFFFAOYSA-N
SMILESFC1=C(Br)C=C(OC(F)(F)F)C=C1
さらに表示
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4475-1.4515 @ 20?C
1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene used as solvents. Mostly employed in chemical processing, extraction and separation of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene used as solvents. Mostly employed in chemical processing, extraction and separation of chemicals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
R.A. Yadav,; I.S. Singh.Vibrational studies of trifluoromethyl benzene derivatives—III. p-trifluoromethyl aniline. Spectrochimica Acta Part A: Molecular Spectroscopy. 1985, 41 (1-2), 191-197.