2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride, 95%, Thermo Scientific Chemicals
2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride, 95%, Thermo Scientific Chemicals
2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride, 95%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
H32717.03
または、製品番号H32717-03
1 g
製品番号(カタログ番号) H32717.03
または、製品番号H32717-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1138011-22-9
IUPAC Name2,2,2-trifluoro-1-(pyridin-3-yl)ethan-1-amine hydrochloride
Molecular FormulaC7H8ClF3N2
InChI KeyNLDVAGWIUPSGQC-UHFFFAOYNA-N
SMILESCl.NC(C1=CN=CC=C1)C(F)(F)F
さらに表示
Appearance (Color)White
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only