3-Methylbenzoic anhydride, 97%
3-Methylbenzoic anhydride, 97%
3-Methylbenzoic anhydride, 97%
3-Methylbenzoic anhydride, 97%
Thermo Scientific Chemicals

3-Methylbenzoic anhydride, 97%

CAS: 21436-44-2 | C16H14O3 | 254.285 g/mol
製品番号(カタログ番号) H32739.03
または、製品番号H32739-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS21436-44-2
IUPAC Name3-methylbenzoyl 3-methylbenzoate
Molecular FormulaC16H14O3
InChI KeyDGZSQLHPEZFGSI-UHFFFAOYSA-N
SMILESCC1=CC(=CC=C1)C(=O)OC(=O)C1=CC(C)=CC=C1
さらに表示
Appearance (Color)White
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder or flakes
Melting Point (clear melt)65.5-74.5°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only