Boron trichloride, 1M soln. in dichloromethane, stab.
Boron trichloride, 1M soln. in dichloromethane, stab.
Boron trichloride, 1M soln. in dichloromethane, stab.
Boron trichloride, 1M soln. in dichloromethane, stab.
Thermo Scientific Chemicals

Boron trichloride, 1M soln. in dichloromethane, stab.

CAS: 10294-34-5 | BCl3 | 117.16 g/mol
数量:
100 mL
500 mL
製品番号(カタログ番号) H32751.AP
または、製品番号H32751-AP
価格(JPY)
-
見積もりを依頼する
数量:
500 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10294-34-5
IUPAC Nametrichloroborane
Molecular FormulaBCl3
InChI KeyFAQYAMRNWDIXMY-UHFFFAOYSA-N
SMILESClB(Cl)Cl
さらに表示
Assay (Titration ex Chloride)0.90-1.05M
FormLiquid
CommentMay form precipitate on storage
Appearance (Color)Clear colorless to pale yellow
Boron trichloride is used in semiconductor production and in the preparation of other boron compounds. It serves as a reagent in the preparation of various protease inhibitors. It is also used as a catalyst and soldering flux. It is involved in the improvement of aluminum castings by treating with boron trichloride vapors. It is utilized to produce electrical resistors. It is also used as a metal refining agent, especially for aluminum, magnesium, zinc and copper alloys. Further, it is used in the preparation of nanomaterials such as nanofibers and nanotubes. In addition to this, it is used in the field of high energy fuels and rocket propellants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boron trichloride is used in semiconductor production and in the preparation of other boron compounds. It serves as a reagent in the preparation of various protease inhibitors. It is also used as a catalyst and soldering flux. It is involved in the improvement of aluminum castings by treating with boron trichloride vapors. It is utilized to produce electrical resistors. It is also used as a metal refining agent, especially for aluminum, magnesium, zinc and copper alloys. Further, it is used in the preparation of nanomaterials such as nanofibers and nanotubes. In addition to this, it is used in the field of high energy fuels and rocket propellants.

Solubility
Miscible with dichloromethane, ethanol, carbon tetrachloride, diethyl ether, dimethyl formamide, aromatic solvents, saturated and halogenated hydrocarbon.

Notes
Moisture sensitive. Incompatible with water, oxidizing agents, bases and metals. Store in a cool place.
RUO – Research Use Only

General References:

  1. Rad, A. S.; Shadravan, A.; Soleymani, A. A.; Motaghedi, N. Lewis acid-base surface interaction of some boron compounds with N-doped graphene; first principles study. Curr. Appl. Phys. 2015, 15 (10), 1271-1277.
  2. Khusainova, L. I.; Khafizova, L. O.; Tyumkina, T. V.; Dzhemilev, U. M. Synthesis of halogen-substituted borolanes and 2,3-dihydro-1H-boroles by reactions of aluminacarbocycles with boron trichloride and boron tribromide. Russ. J. Org. Chem. 2014, 50 (3), 309-313.