Methyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate, 98%
Methyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate, 98%
Methyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate, 98%
Methyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate, 98%
Thermo Scientific Chemicals

Methyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate, 98%

CAS: 108354-78-5 | C10H13NO2S | 211.279 g/mol
製品番号(カタログ番号) H32755.14
または、製品番号H32755-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS108354-78-5
IUPAC Namemethyl 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Molecular FormulaC10H13NO2S
InChI KeyDKYYKIHEIOOWRB-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(N)SC2=C1CCCC2
さらに表示
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Appearance (Color)Pale cream to cream to yellow
Assay (GC)≥97.5%
Melting Point (clear melt)128.0-134.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only