Thermo Scientific Chemicals

Dimethyl trisulfide, 98%, Thermo Scientific Chemicals

Catalog number: H32913.14
25 g, Each
Thermo Scientific Chemicals

Dimethyl trisulfide, 98%, Thermo Scientific Chemicals

Catalog number: H32913.14
25 g, Each
Quantity
Catalog number: H32913.14
also known as H32913-14
Price (USD)
Price: 360.00
Online price: 311.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3658-80-8
IUPAC Name
dimethyltrisulfane
Molecular Formula
C2H6S3
InChI Key
YWHLKYXPLRWGSE-UHFFFAOYSA-N
SMILES
CSSSC
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.5980-1.6020 @ 20?C

Description

Dimethyl trisulfide is reported as a major aroma component in cooked Brassicaceous vegetables. Trap baits containing dimethyl trisulfide have been used to capture Calliphora loewi and other blowflies. Dimethyl trisulfide along with dimethyl sulfide and dimethyl disulfide have been confirmed as volatile compounds given off by the fly-attracting plant known as dead-horse arum (Helicodiceros muscivorus). These flies are attracted to the odor of fetid meat and help pollinate this plant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl trisulfide is reported as a major aroma component in cooked Brassicaceous vegetables. Trap baits containing dimethyl trisulfide have been used to capture Calliphora loewi and other blowflies. Dimethyl trisulfide along with dimethyl sulfide and dimethyl disulfide have been confirmed as volatile compounds given off by the fly-attracting plant known as dead-horse arum (Helicodiceros muscivorus). These flies are attracted to the odor of fetid meat and help pollinate this plant.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C. Store in an air-tight and light-resistant container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text