Ethyl 2-fluoroisobutyrate, 97%, Thermo Scientific Chemicals
Ethyl 2-fluoroisobutyrate, 97%, Thermo Scientific Chemicals
Ethyl 2-fluoroisobutyrate, 97%, Thermo Scientific Chemicals
Ethyl 2-fluoroisobutyrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-fluoroisobutyrate, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
H32931.06
または、製品番号H32931-06
5 g
製品番号(カタログ番号) H32931.06
または、製品番号H32931-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS55816-69-8
IUPAC Nameethyl 2-fluoro-2-methylpropanoate
Molecular FormulaC6H11FO2
InChI KeyCJRQQJKWNULSFQ-UHFFFAOYSA-N
SMILESCCOC(=O)C(C)(C)F
さらに表示
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.3770-1.3820 @ 20?C
Ethyl 2-fluoroisobutyrate is used as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-fluoroisobutyrate is used as an active pharmaceutical ingredient.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
Gauri S. Lal.; Guido P. Pez.; Reno J. Pesaresi.; Frank M. Prozonic.; Hansong Cheng. Bis(2-methoxyethyl)aminosulfur Trifluoride:  A New Broad-Spectrum Deoxofluorinating Agent with Enhanced Thermal Stability. J. Org. Chem. 1999, 64 (19), 7048-7054.