4-Bromo-2,3,5,6-tetrafluorobenzoic acid is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromo-2,3,5,6-tetrafluorobenzoic acid is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
RUO – Research Use Only
Christ Tamborski.; Edward J. Soloski. Organolithium Intermediates from 1,4-Disubstituted Tetrafluorobenzene Compounds. J. Org. Chem. 1966, 31 (3), 743-745.