4-(2-Benzothiazolyl)aniline, 98%
4-(2-Benzothiazolyl)aniline, 98%
4-(2-Benzothiazolyl)aniline, 98%
Thermo Scientific Chemicals

4-(2-Benzothiazolyl)aniline, 98%

CAS: 6278-73-5 | C13H10N2S | 226.30 g/mol
製品番号(カタログ番号) H33052.03
または、製品番号H33052-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6278-73-5
IUPAC Name4-(1,3-benzothiazol-2-yl)aniline
Molecular FormulaC13H10N2S
InChI KeyWKRCOZSCENDENK-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)C1=NC2=CC=CC=C2S1
さらに表示
Appearance (Color)Cream to yellow
Assay (GC)≥97.5%
FormPowder
Melting Point (clear melt)151.0-158.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only