Quinoxaline-6-carboxylic acid, 95%
Quinoxaline-6-carboxylic acid, 95%
Quinoxaline-6-carboxylic acid, 95%
Quinoxaline-6-carboxylic acid, 95%
Thermo Scientific Chemicals

Quinoxaline-6-carboxylic acid, 95%

CAS: 6925-00-4 | C9H6N2O2 | 174.159 g/mol
製品番号(カタログ番号) H33157.03
または、製品番号H33157-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6925-00-4
IUPAC Namequinoxaline-6-carboxylate
Molecular FormulaC9H5N2O2
InChI KeyJGQDBVXRYDEWGM-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC=C2N=CC=NC2=C1
さらに表示
Appearance (Color)Grey to dark grey
FormPowder
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
Assay (Silylated GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only