11-Azidoundecyltriethoxysilane, 95%
11-Azidoundecyltriethoxysilane, 95%
11-Azidoundecyltriethoxysilane, 95%
Thermo Scientific Chemicals

11-Azidoundecyltriethoxysilane, 95%

CAS: 663171-33-3 | C17H37N3O3Si | 359.586 g/mol
製品番号(カタログ番号) H33524.MD
または、製品番号H33524-MD
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS663171-33-3
IUPAC Name(11-azidoundecyl)triethoxysilane
Molecular FormulaC17H37N3O3Si
InChI KeyWCDFLYKXACDAES-UHFFFAOYSA-N
SMILESCCO[Si](CCCCCCCCCCCN=[N+]=[N-])(OCC)OCC
さらに表示
FormLiquid
Refractive Index1.4400-1.4460 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥94.0%
This coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Jef Ryken,; Jiaqi Li,; Tim Steylaerts,; Rita Vos,; Josine Loo,; Karolien Jans,; Willem Van Roy,; Tim Stakenborg,; Pol Van Dorpe,; Jeroen Lammertyn,; Liesbet Lagae. Biosensing with SiO2-covered SPR substrates in a commercial SPR-tool . Sensors and Actuators B: Chemical. 2014, 200 167-172.
  2. Jonathan Sabaté del Río,; Tim Steylaerts,; Olivier Y.F. Henry,; Peter Bienstman,; Tim Stakenborg,; Wim Van Roy,; Ciara K. O’Sullivan. Real-time and label-free ring-resonator monitoring of solid-phase recombinase polymerase amplification. Biosensors and Bioelectronics. 2015, 73 130-137.