Thermo Scientific Chemicals

2-Fluoro-4-(trifluoromethoxy)benzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: H33749.06
5 g, Each
Thermo Scientific Chemicals

2-Fluoro-4-(trifluoromethoxy)benzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: H33749.06
5 g, Each
Quantity
Catalog number: H33749.06
also known as H33749-06
Price (USD)

Chemical Identifiers

CAS
1073477-22-1
Melting Point (clear melt)
131.0-137.0?C
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥96.0%
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Appearance (Color)
White

Description

2-Fluoro-4-(trifluoromethyl)benzoic acid may be used in pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-4-(trifluoromethyl)benzoic acid may be used in pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text