1-Chloromethyl-1H-1,2,4-triazole hydrochloride, 96%
Thermo Scientific Chemicals

1-Chloromethyl-1H-1,2,4-triazole hydrochloride, 96%

CAS: 84387-61-1 | C3H4ClN3 | 117.54 g/mol
製品番号(カタログ番号) H33830.03
または、製品番号H33830-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS84387-61-1
Appearance (Color)White to cream or pale yellow to yellow
FormPowder or crystalline powder
Assay (Titration ex Chloride)≥95.0% to ≤105.0%
Water Content (Karl Fischer Titration)≤3.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only