n-Heptane, 95%
n-Heptane, 95%
n-Heptane, 95%
Thermo Scientific Chemicals

n-Heptane, 95%

CAS: 142-82-5 | C7H16 | 100.21 g/mol
数量:
1000 mL
2500 mL
製品番号(カタログ番号) H33987.AU
または、製品番号H33987-AU
価格(JPY)
-
数量:
1000 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS142-82-5
IUPAC Nameheptane
Molecular FormulaC7H16
InChI KeyIMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILESCCCCCCC
さらに表示
Assay (GC)≥94.0%
Identification (FTIR)Conforms
Refractive Index1.3850-1.3910 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
n-Heptane is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Heptane is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.

Solubility
Miscible with alcohol, chloroform, and ether. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area away from incompatible substances.
RUO – Research Use Only

General References:

  1. Caponetti, E.; Pedone, L.; Martino, D. C.; Panto, V.; Liveri, V. T. Synthesis, size control, and passivation of CdS nanoparticles in water/AOT-heptane microemulsions. Mater. Sci. Eng. C 2003, 23 (4), 531-539.
  2. Chica, A.; Corma, A. Hydroisomerization of Pentane, Hexane, and Heptane for Improving the Octane Number of Gasoline. J. Catal. 1999, 187 (1), 167-176.