4'-Methylbiphenyl-4-carboxaldehyde, 96%
4'-Methylbiphenyl-4-carboxaldehyde, 96%
4'-Methylbiphenyl-4-carboxaldehyde, 96%
Thermo Scientific Chemicals

4'-Methylbiphenyl-4-carboxaldehyde, 96%

CAS: 36393-42-7 | C14H12O | 196.249 g/mol
製品番号(カタログ番号) H34249.MD
または、製品番号H34249-MD
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS36393-42-7
IUPAC Name4'-methyl-[1,1'-biphenyl]-4-carbaldehyde
Molecular FormulaC14H12O
InChI KeyBCINBWXQYBLSKO-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C1=CC=C(C=O)C=C1
さらに表示
Assay (GC)≥95.0%
Appearance (Color)White
FormPowder
Free acid (titration)≤2%
Melting Point (clear melt)102.0-112.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only