4-(2-Thienyl)benzenesulfonyl chloride, 96%
4-(2-Thienyl)benzenesulfonyl chloride, 96%
4-(2-Thienyl)benzenesulfonyl chloride, 96%
Thermo Scientific Chemicals

4-(2-Thienyl)benzenesulfonyl chloride, 96%

CAS: 209592-91-6 | C10H7ClO2S2 | 258.734 g/mol
数量:
250 mg
1 g
5 g
製品番号(カタログ番号) H34399.03
または、製品番号H34399-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS209592-91-6
IUPAC Name4-(thiophen-2-yl)benzene-1-sulfonyl chloride
Molecular FormulaC10H7ClO2S2
InChI KeyIRDZCMUQNGEQEP-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1=CC=C(C=C1)C1=CC=CS1
さらに表示
Assay (Titration ex Chloride)≥95.0 to ≤105.0%
Appearance (Color)Pale yellow
FormCrystals or powder or crystalline powder
Melting Point (clear melt)109.5-118.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only