4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride, 95%
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride, 95%
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride, 95%
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride, 95%
Thermo Scientific Chemicals

4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride, 95%

CAS: 128852-17-5 | C8H9ClO2S2 | 236.73 g/mol
製品番号(カタログ番号) H34470.MD
または、製品番号H34470-MD
価格(JPY)
-
見積もりを依頼する
数量:
250 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS128852-17-5
IUPAC Name4,5,6,7-tetrahydro-1-benzothiophene-2-sulfonyl chloride
Molecular FormulaC8H9ClO2S2
InChI KeyUUBSHLWNADKCBU-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1=CC2=C(CCCC2)S1
さらに表示
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥94.0%
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
Melting Point (clear melt)57.5-69.5?C
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water

Notes
Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only