2-Bromo-6-iodopyridine, 97%
2-Bromo-6-iodopyridine, 97%
2-Bromo-6-iodopyridine, 97%
Thermo Scientific Chemicals

2-Bromo-6-iodopyridine, 97%

CAS: 234111-08-1 | C5H3BrIN | 283.89 g/mol
製品番号(カタログ番号) H34478.03
または、製品番号H34478-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS234111-08-1
Appearance (Color)White to cream to brown
Assay (GC)≥96.0%
Melting Point (clear melt)137.0-147.0?C
FormCrystals or powder or crystalline powder

6-trifluoromethyl-2-pyridinecarboxylic acid (4; 87%) was obtained from 2-bromo-6-(trifluoromethyl)pyridine, which was in turn derived from 2-bromo-6-iodopyridine by trifluoromethylating displacement of iodine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-trifluoromethyl-2-pyridinecarboxylic acid (4; 87%) was obtained from 2-bromo-6-(trifluoromethyl)pyridine, which was in turn derived from 2-bromo-6-iodopyridine by trifluoromethylating displacement of iodine.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Fabrice Cottet, Marc Marull1, Olivier Lefebvre1 andManfred Schlosser.Recommendable Routes to Trifluoromethyl-Substituted Pyridine- and Quinolinecarboxylic Acids. European Journal of Organic Chemistry. 2003, 8 1559-1568.
  2. Tatsushi Imahori,; Masanobu Uchiyama,; Takao Sakamoto,; Yoshinori Kondo. Regiocontrolled deprotonative-zincation of bromopyridines using aminozincates. Chem. Commun., 2001, (23), 2450-2451.