6-Fluoro-1,2,3,4-tetrahydroquinoline, 97%
6-Fluoro-1,2,3,4-tetrahydroquinoline, 97%
6-Fluoro-1,2,3,4-tetrahydroquinoline, 97%
Thermo Scientific Chemicals

6-Fluoro-1,2,3,4-tetrahydroquinoline, 97%

CAS: 59611-52-8 | C9H10FN | 151.18 g/mol
製品番号(カタログ番号) H35364.03
または、製品番号H35364-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS59611-52-8
IUPAC Name6-fluoro-1,2,3,4-tetrahydroquinoline
Molecular FormulaC9H10FN
InChI KeyNECDDBBJVCNJNS-UHFFFAOYSA-N
SMILESFC1=CC2=C(NCCC2)C=C1
さらに表示
Appearance (Color)White to pale cream or pale yellow to yellow
FormFused solid or crystalline powder and/or needles
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)26.0-36.0°C
6-Fluoro-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Fluoro-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Jozsef Balint; Gabriella Egri; Violetta Kiss; Antal Gajary; Zoltan Juvancz; Elemer Fogassy. Unusual phenomena during the resolution of 6-fluoro-2-methyl-1,2,3,4-tetrahydroquinoline (FTHQ): thermodynamic-kinetic control. Tetrahedron: Asymmetry. 2002, 12, (24),3435-3439