Thermo Scientific Chemicals

4-Aminonicotinic acid, 98%, Thermo Scientific Chemicals

Catalog number: H50032.06
5 g, Each
Thermo Scientific Chemicals

4-Aminonicotinic acid, 98%, Thermo Scientific Chemicals

Catalog number: H50032.06
5 g, Each
Quantity
Catalog number: H50032.06
also known as H50032-06
Price (USD)
Price: 92.70
Online price: 79.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7418-65-7
IUPAC Name
4-aminopyridine-3-carboxylic acid
Molecular Formula
C6H6N2O2
InChI Key
IASBMUIXBJNMDW-UHFFFAOYSA-N
SMILES
NC1=CC=NC=C1C(O)=O
Assay (unspecified)
97.5% min
Appearance (Color)
White to yellow
Form
Solid

Description

4-Aminonicotinic acid has been used in the preparation of 2-methyl-pyrido-oxazine. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminonicotinic acid has been used in the preparation of 2-methyl-pyrido-oxazine. It is also used as pharmaceutical intermediate.

Solubility
Soluble in aqueous acid and base.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text